Product Details

SNP ID
rs75152309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40876571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCATCACTCTTCTTAAACTGCCCC[A/T]TGTCATCCAGGAAATGCTGGGGATT
Phenotype
MIM: 608054
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CYP2A7 PubMed Links

Gene Details

Gene
CYP2A7
Gene Name
cytochrome P450 family 2 subfamily A member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000764.2 Intron NP_000755.2
NM_030589.2 Intron NP_085079.2
XM_017026383.1 Intron XP_016881872.1

View Full Product Details