Product Details

SNP ID
rs74864344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41364687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACTTCCTTCTCGGAGCTACTAC[C/T]AAGTTCACTGTTACTGCTTGATTCT
Phenotype
MIM: 611951
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B9D2 PubMed Links

Gene Details

Gene
B9D2
Gene Name
B9 protein domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030578.3 Intron NP_085055.2
XM_011527349.2 Intron XP_011525651.1
XM_011527350.1 Intron XP_011525652.1
Gene
TMEM91
Gene Name
transmembrane protein 91
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042595.2 Intron NP_001036060.1
NM_001098821.1 Intron NP_001092291.1
NM_001098822.1 Intron NP_001092292.1
NM_001098823.1 Intron NP_001092293.1
NM_001098824.1 Intron NP_001092294.1
NM_001098825.1 Intron NP_001092295.1

View Full Product Details