Product Details

SNP ID
rs78739853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43612954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCAGGCCAGCACCCGCAGCAGGC[C/T]GCAAAGTCACAGCCAATCTAGAAGC
Phenotype
MIM: 609744
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CADM4 PubMed Links

Gene Details

Gene
CADM4
Gene Name
cell adhesion molecule 4
There are no transcripts associated with this gene.

Gene
SRRM5
Gene Name
serine/arginine repetitive matrix 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145641.1 854 Missense Mutation CCG,CTG P278L NP_001139113.1
Gene
ZNF428
Gene Name
zinc finger protein 428
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182498.3 854 Intron NP_872304.2

View Full Product Details