Product Details
- SNP ID
-
rs78718926
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:44476637 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTTGAGTAAGGCATGAACTCTGT[C/T]TGAAGGACTTCCCACATTGACTGCA
- Phenotype
-
MIM: 606740
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF180
PubMed Links
Gene Details
- Gene
- ZNF180
- Gene Name
- zinc finger protein 180
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001278508.2 |
2005 |
Missense Mutation |
AAA,AGA |
K590R |
NP_001265437.2 |
NM_001278509.2 |
2005 |
Missense Mutation |
AAA,AGA |
K588R |
NP_001265438.2 |
NM_001288759.2 |
2005 |
Missense Mutation |
AAA,AGA |
K614R |
NP_001275688.2 |
NM_001288760.2 |
2005 |
Missense Mutation |
AAA,AGA |
K272R |
NP_001275689.1 |
NM_001288761.2 |
2005 |
Missense Mutation |
AAA,AGA |
K272R |
NP_001275690.1 |
NM_001288762.2 |
2005 |
Missense Mutation |
AAA,AGA |
K272R |
NP_001275691.1 |
NM_001291633.1 |
2005 |
Missense Mutation |
AAA,AGA |
K588R |
NP_001278562.1 |
NM_013256.5 |
2005 |
Missense Mutation |
AAA,AGA |
K615R |
NP_037388.3 |
XM_011527280.2 |
2005 |
Missense Mutation |
AAA,AGA |
K657R |
XP_011525582.1 |
XM_017027250.1 |
2005 |
Missense Mutation |
AAA,AGA |
K272R |
XP_016882739.1 |
XM_017027251.1 |
2005 |
Missense Mutation |
AAA,AGA |
K272R |
XP_016882740.1 |
XM_017027252.1 |
2005 |
Missense Mutation |
AAA,AGA |
K272R |
XP_016882741.1 |
XM_017027253.1 |
2005 |
Missense Mutation |
AAA,AGA |
K272R |
XP_016882742.1 |
XM_017027254.1 |
2005 |
Missense Mutation |
AAA,AGA |
K272R |
XP_016882743.1 |
XM_017027255.1 |
2005 |
Missense Mutation |
AAA,AGA |
K272R |
XP_016882744.1 |
XM_017027256.1 |
2005 |
Missense Mutation |
AAA,AGA |
K272R |
XP_016882745.1 |
View Full Product Details