Product Details

SNP ID
rs78276697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46720645 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGCATCAGCGCCAGCACTGGCA[A/C]TGAGGGCCTTGCTGGGGTGGCAGGC
Phenotype
MIM: 607074 MIM: 614767
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC107987269 PubMed Links

Gene Details

Gene
LOC107987269
Gene Name
uncharacterized LOC107987269
There are no transcripts associated with this gene.

Gene
PRKD2
Gene Name
protein kinase D2
There are no transcripts associated with this gene.

Gene
STRN4
Gene Name
striatin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039877.1 2309 Missense Mutation AGT,ATT S747I NP_001034966.1
NM_013403.2 2309 Missense Mutation AGT,ATT S740I NP_037535.2
XM_006723171.2 2309 Missense Mutation AGT,ATT S762I XP_006723234.1
XM_006723172.2 2309 Missense Mutation AGT,ATT S755I XP_006723235.1
XM_006723174.2 2309 Missense Mutation AGT,ATT S628I XP_006723237.1
XM_011526878.1 2309 UTR 3 XP_011525180.1
XM_011526879.1 2309 Missense Mutation AGT,ATT S352I XP_011525181.1
XM_017026716.1 2309 Missense Mutation AGT,ATT S643I XP_016882205.1
XM_017026717.1 2309 Missense Mutation AGT,ATT S628I XP_016882206.1
XM_017026718.1 2309 Missense Mutation AGT,ATT S526I XP_016882207.1
XM_017026719.1 2309 UTR 3 XP_016882208.1

View Full Product Details