Product Details

SNP ID
rs77520296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48404830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGAGTATGACTGGACGTCCTTT[G/T]TAGCCGTGACCACTCGTGCCCCTGG
Phenotype
MIM: 602717
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GRIN2D PubMed Links

Gene Details

Gene
GRIN2D
Gene Name
glutamate ionotropic receptor NMDA type subunit 2D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000836.2 650 Missense Mutation GTA,TTA V188L NP_000827.2
XM_011526872.1 650 Missense Mutation GTA,TTA V188L XP_011525174.1

View Full Product Details