Product Details

SNP ID
rs75776802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49514339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACCTCAAGCAGGGCACCTGGGGT[G/T]GGGACTGGCCCGAGGCCCTGGCTAT
Phenotype
MIM: 601437
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FCGRT PubMed Links

Gene Details

Gene
FCGRT
Gene Name
Fc fragment of IgG receptor and transporter
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136019.2 676 Missense Mutation GGG,TGG G152W NP_001129491.1
NM_004107.4 676 Missense Mutation GGG,TGG G152W NP_004098.1
XM_005258657.1 676 Missense Mutation GGG,TGG G12W XP_005258714.1
XM_011526627.2 676 Intron XP_011524929.1

View Full Product Details