Product Details

SNP ID
rs77869147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49536964 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTACCCCAGTAGGATTTACTTT[G/T]TCTTTGTTTTAACCTGCTTGGCGTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RCN3 PubMed Links

Gene Details

Gene
RCN3
Gene Name
reticulocalbin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020650.2 Intron NP_065701.2
XM_005259089.3 Intron XP_005259146.1
XM_011527143.1 Intron XP_011525445.1
XM_017027023.1 Intron XP_016882512.1

View Full Product Details