Product Details

SNP ID
rs78405916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49852583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTAACCGTCGTTTTTAAATATGA[C/T]CGTCCCATCTTGATGTGCTGCTCCT
Phenotype
MIM: 605610 MIM: 610195
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4749 PubMed Links

Gene Details

Gene
MIR4749
Gene Name
microRNA 4749
There are no transcripts associated with this gene.

Gene
PNKP
Gene Name
polynucleotide kinase 3'-phosphatase
There are no transcripts associated with this gene.

Gene
PTOV1
Gene Name
prostate tumor overexpressed 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305105.1 Intron NP_001292034.1
NM_001305108.1 Intron NP_001292037.1
NM_017432.4 Intron NP_059128.2
XM_005258998.4 Intron XP_005259055.1
XM_006723244.3 Intron XP_006723307.1
XM_011527033.1 Intron XP_011525335.1
XM_011527034.2 Intron XP_011525336.1
XM_011527035.1 Intron XP_011525337.1
XM_017026879.1 Intron XP_016882368.1
XM_017026880.1 Intron XP_016882369.1
Gene
PTOV1-AS1
Gene Name
PTOV1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
PTOV1-AS2
Gene Name
PTOV1 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details