Product Details

SNP ID
rs77528293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50469221 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATGAACACACCAGGCTCAGTCC[A/G]ACCCTAGCACCTTTGCACAGATGGT
Phenotype
MIM: 614545 MIM: 160793
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EMC10 PubMed Links

Gene Details

Gene
EMC10
Gene Name
ER membrane protein complex subunit 10
There are no transcripts associated with this gene.

Gene
FAM71E1
Gene Name
family with sequence similarity 71 member E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308429.1 Intron NP_001295358.1
NM_138411.2 Intron NP_612420.1
XM_011526405.2 Intron XP_011524707.2
XM_011526406.2 Intron XP_011524708.2
Gene
MYBPC2
Gene Name
myosin binding protein C, fast type
There are no transcripts associated with this gene.

View Full Product Details