Product Details

SNP ID
rs74443808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51104013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCCGCCTCCTCGCCTGTGCATT[A/C]GCCCCACTGCCTCGGAGAGTGCCGC
Phenotype
MIM: 612694
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CTU1 PubMed Links

Gene Details

Gene
CTU1
Gene Name
cytosolic thiouridylase subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145232.3 Intron NP_660275.2

View Full Product Details