Product Details

SNP ID
rs76377651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51453850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTAGTGGCGTAAGGATGACAAGCC[A/G]GTCAAAGAAATCAGGTCCCGCCAAT
Phenotype
MIM: 605639
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIGLEC8 PubMed Links

Gene Details

Gene
SIGLEC8
Gene Name
sialic acid binding Ig like lectin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014442.2 Intron NP_055257.2
XM_011526734.2 Intron XP_011525036.1
XM_011526735.2 Intron XP_011525037.1

View Full Product Details