Product Details

SNP ID
rs75588884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51478692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTGTGGAGGAGGGTCTTCCTCA[C/T]GGGTAAGAGATGCTGGATGCTGGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEACAM18 PubMed Links

Gene Details

Gene
CEACAM18
Gene Name
carcinoembryonic antigen related cell adhesion molecule 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278392.1 50 Missense Mutation ACG,ATG T17M NP_001265321.1
XM_011527258.2 50 Missense Mutation ACG,ATG T17M XP_011525560.1

View Full Product Details