Product Details

SNP ID
rs75554203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55093207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCTTCCCTGCAGCCTTCTCTGC[C/T]TCCTTCAGGTCTGTAAGAGTCACAC
Phenotype
MIM: 614987 MIM: 613245
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPS8L1 PubMed Links

Gene Details

Gene
EPS8L1
Gene Name
EPS8 like 1
There are no transcripts associated with this gene.

Gene
PPP1R12C
Gene Name
protein phosphatase 1 regulatory subunit 12C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271618.1 1756 Silent Mutation GAA,GAG E569E NP_001258547.1
NM_017607.3 1756 Silent Mutation GAA,GAG E570E NP_060077.1
XM_005259013.3 1756 Silent Mutation GAA,GAG E570E XP_005259070.1
XM_011527045.2 1756 Silent Mutation GAA,GAG E569E XP_011525347.1

View Full Product Details