Product Details

SNP ID
rs77942970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55947936 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGAATCCACCCTCTGACACCCCC[A/C]TTCCCTTTTCATCCTCCTCCACTCA
Phenotype
MIM: 609659
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NLRP8 PubMed Links

Gene Details

Gene
NLRP8
Gene Name
NLR family pyrin domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317000.1 105 Missense Mutation ATT,CTT I12L NP_001303929.1
NM_176811.2 105 Missense Mutation ATT,CTT I12L NP_789781.2

View Full Product Details