Product Details

SNP ID
rs77606741
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56143630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGCTCCTGCTGCAACTTGGGGG[C/T]AGTAACACCAACCTCATGGGATTGA
Phenotype
MIM: 607874
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF444 PubMed Links

Gene Details

Gene
ZNF444
Gene Name
zinc finger protein 444
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253792.1 Intron NP_001240721.1
NM_018337.3 Intron NP_060807.2
XM_005259034.2 Intron XP_005259091.1
XM_005259035.1 Intron XP_005259092.1
XM_005259036.1 Intron XP_005259093.1
XM_011527068.1 Intron XP_011525370.1
XM_011527069.1 Intron XP_011525371.1
XM_011527070.1 Intron XP_011525372.1
XM_011527071.1 Intron XP_011525373.1
XM_011527073.1 Intron XP_011525375.1
XM_017026932.1 Intron XP_016882421.1
XM_017026933.1 Intron XP_016882422.1

View Full Product Details