Product Details
- SNP ID
-
rs77114013
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:56664728 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGCTTCTCGCCCGTGTGCGTGCG[C/G]TGGTGCAGGGTCAGGTGCACGCTCT
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SMIM17
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs8108756] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SMIM17
- Gene Name
- small integral membrane protein 17
There are no transcripts associated with this gene.
- Gene
- ZNF835
- Gene Name
- zinc finger protein 835
View Full Product Details