Product Details

SNP ID
rs75663143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57618176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGAAAAGGGCTGAGTTTATCTGG[A/G]ATCTGCACATGGTTCTGGATTGCTG
Phenotype
MIM: 604076
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF134 PubMed Links

Gene Details

Gene
ZNF134
Gene Name
zinc finger protein 134
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003435.4 Intron NP_003426.3
Gene
ZNF530
Gene Name
zinc finger protein 530
There are no transcripts associated with this gene.

View Full Product Details