Product Details

SNP ID
rs76439307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57652549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCGATGAACCTCTGCCCTTGAA[A/C]CCCTCCTCACCACCATATCCTCCTC
Phenotype
MIM: 601856 MIM: 613419
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF211 PubMed Links

Gene Details

Gene
ZNF211
Gene Name
zinc finger protein 211
There are no transcripts associated with this gene.

Gene
ZSCAN4
Gene Name
zinc finger and SCAN domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152677.2 Intron NP_689890.1
XM_011526607.2 Intron XP_011524909.1
XM_017026458.1 Intron XP_016881947.1

View Full Product Details