Product Details

SNP ID
rs75294658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57912424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTCCTGACATAAAAAGGTCCAT[C/T]CCCTTCTGAATAGCGATATCCAAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF417 PubMed Links

Gene Details

Gene
ZNF417
Gene Name
zinc finger protein 417
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297734.1 Intron NP_001284663.1
NM_152475.2 Intron NP_689688.2
XM_011526473.2 Intron XP_011524775.1
XM_017026315.1 Intron XP_016881804.1
Gene
ZNF418
Gene Name
zinc finger protein 418
There are no transcripts associated with this gene.

View Full Product Details