Product Details

SNP ID
rs75188336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57923377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGTATTTTTCTATTTAGTGACA[A/C]ACTTACCTAACCCATTTATGCCAAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF417 PubMed Links

Gene Details

Gene
ZNF417
Gene Name
zinc finger protein 417
There are no transcripts associated with this gene.

Gene
ZNF418
Gene Name
zinc finger protein 418
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317027.1 Intron NP_001303956.1
NM_001317028.1 Intron NP_001303957.1
NM_001317029.1 Intron NP_001303958.1
NM_001317030.1 Intron NP_001303959.1
NM_133460.2 Intron NP_597717.1
XM_017026304.1 Intron XP_016881793.1
XM_017026305.1 Intron XP_016881794.1
XM_017026306.1 Intron XP_016881795.1
XM_017026307.1 Intron XP_016881796.1
XM_017026308.1 Intron XP_016881797.1
XM_017026309.1 Intron XP_016881798.1
XM_017026310.1 Intron XP_016881799.1
XM_017026311.1 Intron XP_016881800.1
XM_017026312.1 Intron XP_016881801.1
XM_017026313.1 Intron XP_016881802.1
XM_017026314.1 Intron XP_016881803.1

View Full Product Details