Product Details

SNP ID
rs74560099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58410088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGAACTTTGCACTCGTTAGCTCA[C/G]TGGGTAAGTCTCTTACACTGTCCCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100506634 PubMed Links

Gene Details

Gene
LOC100506634
Gene Name
hepatocellular carcinoma-associated antigen HCA25a
There are no transcripts associated with this gene.

Gene
ZNF584
Gene Name
zinc finger protein 584
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318002.1 640 Missense Mutation CTG,GTG L48V NP_001304931.1
NM_173548.2 640 Missense Mutation CTG,GTG L56V NP_775819.1
XM_005258628.4 640 Missense Mutation CTG,GTG L48V XP_005258685.1
XM_005258630.4 640 Missense Mutation CTG,GTG L56V XP_005258687.1

View Full Product Details