Product Details
- SNP ID
-
rs79180036
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:32836975 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTAATTTTTCACAGCTTTGAAGAT[A/C]CGCCTGAAAAATCTTTTACTTTCAA
- Phenotype
-
MIM: 604144
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC7A9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142034546] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC7A9
- Gene Name
- solute carrier family 7 member 9
- Gene
- TDRD12
- Gene Name
- tudor domain containing 12
There are no transcripts associated with this gene.
View Full Product Details