Product Details

SNP ID
rs74887182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35282773 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAGCACTGAGCAGAGCTCAGGAC[A/G]TCTCAGGAGTACTGGCAGCTGAACA
Phenotype
MIM: 606464 MIM: 159460 MIM: 600390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HAMP PubMed Links

Gene Details

Gene
HAMP
Gene Name
hepcidin antimicrobial peptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021175.3 Intron NP_066998.1
Gene
MAG
Gene Name
myelin associated glycoprotein
There are no transcripts associated with this gene.

Gene
USF2
Gene Name
upstream transcription factor 2, c-fos interacting
There are no transcripts associated with this gene.

View Full Product Details