Product Details

SNP ID
rs77782321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35907512 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTACACGGCCAGGGCAATGAGCA[A/C]TGTCAGCACCAGGTCTCCCATCACG
Phenotype
MIM: 604089 MIM: 604142
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HCST PubMed Links

Gene Details

Gene
HCST
Gene Name
hematopoietic cell signal transducer
There are no transcripts associated with this gene.

Gene
NFKBID
Gene Name
NFKB inhibitor delta
There are no transcripts associated with this gene.

Gene
TYROBP
Gene Name
TYRO protein tyrosine kinase binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173514.1 244 Missense Mutation GTG,TTG V44L NP_001166985.1
NM_001173515.1 244 Missense Mutation GTG,TTG V44L NP_001166986.1
NM_003332.3 244 Missense Mutation GTG,TTG V55L NP_003323.1
NM_198125.2 244 Missense Mutation GTG,TTG V55L NP_937758.1

View Full Product Details