Product Details

SNP ID
rs79014621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36140411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTTCCTTGGGGTGTCCTGAACC[A/G]CGATCGTAGATCTGTGCCCCCAGTT
Phenotype
MIM: 114170
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAPNS1 PubMed Links

Gene Details

Gene
CAPNS1
Gene Name
calpain small subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003962.2 Intron NP_001003962.1
NM_001302632.1 Intron NP_001289561.1
NM_001302633.1 Intron NP_001289562.1
NM_001749.3 Intron NP_001740.1
XM_005259295.1 Intron XP_005259352.1
XM_005259296.1 Intron XP_005259353.1
XM_011527359.1 Intron XP_011525661.1

View Full Product Details