Product Details

SNP ID
rs77092519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36890895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAATAAATTTGACTAACAACTTA[A/T]GTATAAAAGGAAATTACAAATATAT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF345 PubMed Links

Gene Details

Gene
ZNF345
Gene Name
zinc finger protein 345
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242472.1 2261 Intron NP_001229401.1
NM_001242474.1 2261 Intron NP_001229403.1
NM_001242475.1 2261 Intron NP_001229404.1
NM_001242476.1 2261 Intron NP_001229405.1
NM_003419.4 2261 Intron NP_003410.1
XM_011526704.2 2261 Intron XP_011525006.1
XM_017026568.1 2261 Intron XP_016882057.1
XM_017026569.1 2261 Intron XP_016882058.1
XM_017026570.1 2261 Intron XP_016882059.1
XM_017026571.1 2261 Intron XP_016882060.1
XM_017026572.1 2261 Intron XP_016882061.1
XM_017026573.1 2261 Intron XP_016882062.1
Gene
ZNF829
Gene Name
zinc finger protein 829
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037232.3 2261 UTR 3 NP_001032309.2
NM_001171979.1 2261 UTR 3 NP_001165450.1
XM_005258876.4 2261 UTR 3 XP_005258933.1
XM_011526933.2 2261 UTR 3 XP_011525235.1

View Full Product Details