Product Details

SNP ID
rs79175212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1207009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTCATCCACCGCATCGACTCCAC[C/G]GAGGTCATCTACCAGCCGCGCCGCA
Phenotype
MIM: 602216
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
STK11 PubMed Links

Gene Details

Gene
STK11
Gene Name
serine/threonine kinase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000455.4 721 Silent Mutation ACC,ACG T32T NP_000446.1
XM_005259617.2 721 Silent Mutation ACC,ACG T32T XP_005259674.1
XM_005259618.3 721 Silent Mutation ACC,ACG T32T XP_005259675.1
XM_011528209.1 721 UTR 5 XP_011526511.1

View Full Product Details