Product Details

SNP ID
rs75608503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1477665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCCGTAAACCTTTACTTACCAAC[A/G]CTGAAATCTGAATTTCATACCTCTT
Phenotype
MIM: 612034 MIM: 600487
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APC2 PubMed Links

Gene Details

Gene
APC2
Gene Name
APC2, WNT signaling pathway regulator
There are no transcripts associated with this gene.

Gene
C19orf25
Gene Name
chromosome 19 open reading frame 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152482.2 Intron NP_689695.2
XM_005259506.3 Intron XP_005259563.1
XM_006722653.3 Intron XP_006722716.1
XM_017026374.1 Intron XP_016881863.1
Gene
PCSK4
Gene Name
proprotein convertase subtilisin/kexin type 4
There are no transcripts associated with this gene.

View Full Product Details