Product Details

SNP ID
rs77048445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3779162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTGCACAAGCTTCTTGCTCTCCA[G/T]GTACTCCATGCCCTCGGCCACGTGC
Phenotype
MIM: 600038 MIM: 610362
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MATK PubMed Links

Gene Details

Gene
MATK
Gene Name
megakaryocyte-associated tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002378.3 1427 Missense Mutation ATG,CTG M344L NP_002369.2
NM_139354.2 1427 Missense Mutation ATG,CTG M302L NP_647611.1
NM_139355.2 1427 Missense Mutation ATG,CTG M343L NP_647612.1
XM_011528018.1 1427 Missense Mutation ATG,CTG M343L XP_011526320.1
Gene
RAX2
Gene Name
retina and anterior neural fold homeobox 2
There are no transcripts associated with this gene.

View Full Product Details