Product Details

SNP ID
rs79942269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19119720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTTCGGAGACAATGGCCTCGGGA[C/T]CCTCATGCTGCTGGGCCCAGGAGAG
Phenotype
MIM: 610823
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC25A42 PubMed Links

Gene Details

Gene
SLC25A42
Gene Name
solute carrier family 25 member 42
There are no transcripts associated with this gene.

Gene
TMEM161A
Gene Name
transmembrane protein 161A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256766.1 1619 UTR 3 NP_001243695.1
NM_017814.2 1619 UTR 3 NP_060284.1
XM_011528117.2 1619 UTR 3 XP_011526419.1
XM_017026916.1 1619 UTR 3 XP_016882405.1

View Full Product Details