Product Details
- SNP ID
-
rs74349485
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:19711634 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTCTCTCCAGTGTGAGTCCTTTC[A/G]TGCAATCGAATTTGACTGGAACGAA
- Phenotype
-
MIM: 194556
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF14
PubMed Links
Gene Details
- Gene
- ZNF14
- Gene Name
- zinc finger protein 14
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021030.2 |
1801 |
Silent Mutation |
CAC,CAT |
H549H |
NP_066358.2 |
View Full Product Details