Product Details
- SNP ID
-
rs75753298
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:23224068 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACTCCACATACTAGACTCAGATAC[A/G]TATTCTTTACACATCTAACCTACAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF724P
PubMed Links
Gene Details
- Gene
- ZNF724P
- Gene Name
- zinc finger protein 724, pseudogene
There are no transcripts associated with this gene.
View Full Product Details