Product Details

SNP ID
rs76923681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10279106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTTCTCCAAAAAGGTTAGAGGT[G/T]TAATTCCTAGAGGAGGCGACCACTA
Phenotype
MIM: 147840 MIM: 614088
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ICAM1 PubMed Links

Gene Details

Gene
ICAM1
Gene Name
intercellular adhesion molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000201.2 Intron NP_000192.2
Gene
ICAM4
Gene Name
intercellular adhesion molecule 4 (Landsteiner-Wiener blood group)
There are no transcripts associated with this gene.

View Full Product Details