Product Details

SNP ID
rs75264555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10288072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCACTGTGGGCGCTGCGTACCTA[C/T]GCAAGTGCCTAGCTATGAAGTCCCA
Phenotype
MIM: 147840 MIM: 614088 MIM: 601852
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ICAM1 PubMed Links

Gene Details

Gene
ICAM1
Gene Name
intercellular adhesion molecule 1
There are no transcripts associated with this gene.

Gene
ICAM4
Gene Name
intercellular adhesion molecule 4 (Landsteiner-Wiener blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039132.2 977 Missense Mutation ACG,ATG T236M NP_001034221.1
NM_001544.4 977 Missense Mutation CGC,TGC R262C NP_001535.1
NM_022377.3 977 UTR 3 NP_071772.1
Gene
ICAM5
Gene Name
intercellular adhesion molecule 5
There are no transcripts associated with this gene.

View Full Product Details