Product Details

SNP ID
rs78392044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10420824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTCTGTCACTGCCCGGGCCCCG[C/G]GAGGGCCAGGCCACCCTGAAGCCTC
Phenotype
MIM: 600126
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PDE4A PubMed Links

Gene Details

Gene
PDE4A
Gene Name
phosphodiesterase 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111307.1 81 Silent Mutation CGC,CGG R20R NP_001104777.1
NM_001111308.1 81 Intron NP_001104778.1
NM_001111309.1 81 Intron NP_001104779.1
NM_001243121.1 81 Intron NP_001230050.1
NM_006202.2 81 Intron NP_006193.1
XM_005259933.2 81 Intron XP_005259990.1
XM_011528054.1 81 Intron XP_011526356.1
XM_011528055.2 81 Silent Mutation CGC,CGG R20R XP_011526357.1
XM_017026865.1 81 Silent Mutation CGC,CGG R20R XP_016882354.1

View Full Product Details