Product Details

SNP ID
rs79769971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10546995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACGCCGGCACCGGCAGATTGTGT[A/C]CTGGTTCGCCGACCACCCCCGGGCC
Phenotype
MIM: 611340
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATG4D PubMed Links

Gene Details

Gene
ATG4D
Gene Name
autophagy related 4D cysteine peptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281504.1 847 Missense Mutation TAC,TCC Y154S NP_001268433.1
NM_032885.5 847 Missense Mutation TAC,TCC Y217S NP_116274.3
XM_006722927.1 847 Intron XP_006722990.1
Gene
KRI1
Gene Name
KRI1 homolog
There are no transcripts associated with this gene.

Gene
MIR1238
Gene Name
microRNA 1238
There are no transcripts associated with this gene.

View Full Product Details