Product Details

SNP ID
rs74533608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10553439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGATCGTGGGTGTGGCTGGGCGC[A/G]GCGTTCTGAGGGGATGTGGGGTCTG
Phenotype
MIM: 611340
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATG4D PubMed Links

Gene Details

Gene
ATG4D
Gene Name
autophagy related 4D cysteine peptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281504.1 2651 Intron NP_001268433.1
NM_032885.5 2651 Intron NP_116274.3
XM_006722927.1 2651 Intron XP_006722990.1
Gene
KRI1
Gene Name
KRI1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023008.3 2651 UTR 3 NP_075384.3
XM_011528190.2 2651 UTR 3 XP_011526492.1
Gene
MIR1238
Gene Name
microRNA 1238
There are no transcripts associated with this gene.

View Full Product Details