Product Details

SNP ID
rs75607624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11948884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGTAGCAAATGTGATAAAGCAT[G/T]TCATAGTTCTAGTTCCTATCATAGA
Phenotype
MIM: 194543
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF69 PubMed Links

Gene Details

Gene
ZNF69
Gene Name
zinc finger protein 69
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321163.1 1012 Intron NP_001308092.1
NM_021915.3 1012 Intron NP_068734.1
XM_006722873.3 1012 Intron XP_006722936.1
XM_006722874.3 1012 Intron XP_006722937.1
XM_017027231.1 1012 Intron XP_016882720.1
Gene
ZNF700
Gene Name
zinc finger protein 700
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271848.1 1012 Missense Mutation TGT,TTT C290F NP_001258777.1
NM_144566.2 1012 Missense Mutation TGT,TTT C287F NP_653167.1

View Full Product Details