Product Details

SNP ID
rs74729257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12318693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGCATTTATTCGGCCCATCTCC[C/T]GTATGCATTACCATATGTCTTCGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF563 PubMed Links

Gene Details

Gene
ZNF563
Gene Name
zinc finger protein 563
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145276.2 1570 Silent Mutation ACA,ACG T444T NP_660319.1
XM_005259750.4 1570 Intron XP_005259807.1
XM_005259751.3 1570 Intron XP_005259808.1
XM_006722650.3 1570 Intron XP_006722713.1
XM_006722651.3 1570 Intron XP_006722714.1
XM_011527698.1 1570 Intron XP_011526000.1
XM_011527699.2 1570 Silent Mutation ACA,ACG T407T XP_011526001.1
XM_011527700.2 1570 Intron XP_011526002.1
XM_017026332.1 1570 Intron XP_016881821.1

View Full Product Details