Product Details

SNP ID
rs75948786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13738209 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTCTGGGAAGTTTCCTCTTCAT[C/T]GTGGCCTTGGGCCCCAGGGAAATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC130 PubMed Links

Gene Details

Gene
CCDC130
Gene Name
coiled-coil domain containing 130
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320561.1 Intron NP_001307490.1
NM_001320564.1 Intron NP_001307493.1
NM_001320565.1 Intron NP_001307494.1
NM_001320566.1 Intron NP_001307495.1
NM_001320567.1 Intron NP_001307496.1
NM_001320568.1 Intron NP_001307497.1
NM_001320569.1 Intron NP_001307498.1
NM_030818.3 Intron NP_110445.1
XM_005260086.4 Intron XP_005260143.1
XM_011528326.1 Intron XP_011526628.1

View Full Product Details