Product Details

SNP ID
rs75666739
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14478736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTCTCCAGCAGGTCATCCACCTT[C/T]TCAATGGCCTTCTCTTCAAAGGCAG
Phenotype
MIM: 605072 MIM: 601032 MIM: 176802
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GIPC1 PubMed Links

Gene Details

Gene
GIPC1
Gene Name
GIPC PDZ domain containing family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005716.3 711 Silent Mutation GAA,GAG E266E NP_005707.1
NM_202468.2 711 Silent Mutation GAA,GAG E266E NP_974197.1
NM_202469.2 711 Silent Mutation GAA,GAG E169E NP_974198.1
NM_202470.2 711 Silent Mutation GAA,GAG E266E NP_974199.1
NM_202494.2 711 Silent Mutation GAA,GAG E169E NP_974223.1
XM_017026147.1 711 Silent Mutation GAA,GAG E266E XP_016881636.1
XM_017026148.1 711 Silent Mutation GAA,GAG E169E XP_016881637.1
Gene
PKN1
Gene Name
protein kinase N1
There are no transcripts associated with this gene.

Gene
PTGER1
Gene Name
prostaglandin E receptor 1
There are no transcripts associated with this gene.

View Full Product Details