Product Details

SNP ID
rs78719569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15110591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGTATGCCAAGCTGACCCTGTC[A/G]GAGCAGCAGGAAGCCCCTGCCACAG
Phenotype
MIM: 605770
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ILVBL PubMed Links

Gene Details

Gene
ILVBL
Gene Name
ilvB acetolactate synthase like
There are no transcripts associated with this gene.

Gene
SYDE1
Gene Name
synapse defective Rho GTPase homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300910.1 1216 Silent Mutation TCA,TCG S315S NP_001287839.1
NM_033025.5 1216 Silent Mutation TCA,TCG S382S NP_149014.3

View Full Product Details