Product Details

SNP ID
rs79447691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15160763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAACAGACAAGGGAAGTGGCTGGG[C/G]AGGCAGTGCCCCAGTGGTGGTGGCC
Phenotype
MIM: 600276
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NOTCH3 PubMed Links

Gene Details

Gene
NOTCH3
Gene Name
notch 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000435.2 6724 Missense Mutation CCC,GCC P2289A NP_000426.2
XM_005259924.4 6724 Missense Mutation CCC,GCC P2237A XP_005259981.1

View Full Product Details