Product Details

SNP ID
rs80306728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17235885 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCGCGCCGCCCCCTGCGCCGAAG[C/T]GTCCGGCCGCCGCAGGGTAGGGCTC
Phenotype
MIM: 132880
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NR2F6 PubMed Links

Gene Details

Gene
NR2F6
Gene Name
nuclear receptor subfamily 2 group F member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005234.3 676 Missense Mutation CAC,CGC H185R NP_005225.2
Gene
OCEL1
Gene Name
occludin/ELL domain containing 1
There are no transcripts associated with this gene.

View Full Product Details