Product Details

SNP ID
rs78265621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17309687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGGCCTCCAGGCCCCCACTCCC[A/C]CTCTGCTAGACAAAATGGCGCTGTG
Phenotype
MIM: 611840
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ABHD8 PubMed Links

Gene Details

Gene
ABHD8
Gene Name
abhydrolase domain containing 8
There are no transcripts associated with this gene.

Gene
DDA1
Gene Name
DET1 and DDB1 associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024050.5 Intron NP_076955.1
Gene
MRPL34
Gene Name
mitochondrial ribosomal protein L34
There are no transcripts associated with this gene.

View Full Product Details