Product Details

SNP ID
rs80283334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17351506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCAACATTTCAACGGGCCACAG[C/G]CAGGGCTGGGAGTTGGGGGAGAAGG
Phenotype
MIM: 608536 MIM: 607647
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GTPBP3 PubMed Links

Gene Details

Gene
GTPBP3
Gene Name
GTP binding protein 3 (mitochondrial)
There are no transcripts associated with this gene.

Gene
PLVAP
Gene Name
plasmalemma vesicle associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031310.2 2246 UTR 3 NP_112600.1

View Full Product Details