Product Details

SNP ID
rs77850663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17969639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACACGGCTGAATTTGGCCCATGC[A/G]TCGGGTGGCTGAGTCACATCCCCAA
Phenotype
MIM: 602982
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNN1 PubMed Links

Gene Details

Gene
KCNN1
Gene Name
potassium calcium-activated channel subfamily N member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002248.4 Intron NP_002239.2
XM_005259905.1 Intron XP_005259962.1
XM_005259906.1 Intron XP_005259963.1
XM_005259907.2 Intron XP_005259964.1
XM_011528004.1 Intron XP_011526306.1
XM_011528005.2 Intron XP_011526307.1

View Full Product Details