Product Details

SNP ID
rs79029805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45479772 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCAGGAGGAGGGATTTGCAGGT[C/G]ATCTTCCTGGGCCTCCTGAGAGCAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
WFDC2 PubMed Links

Gene Details

Gene
WFDC2
Gene Name
WAP four-disulfide core domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006103.3 Intron NP_006094.3

View Full Product Details