Product Details

SNP ID
rs79792569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:49046950 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTAGTGAAGCCCCCGAACTCAC[A/C]CTTCTCTAACCTCTGTACTTGTATC
Phenotype
MIM: 601342
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CSE1L PubMed Links

Gene Details

Gene
CSE1L
Gene Name
chromosome segregation 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256135.1 Intron NP_001243064.1
NM_001316.3 Intron NP_001307.2
XM_011528599.2 Intron XP_011526901.1
Gene
CSE1L-AS1
Gene Name
CSE1L antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details